Drukuj

Anti-LPIN1 Mouse Monoclonal Antibody [clone: 5H2]

Dostawca: OriGene
ORIGCF806419EA 3950 PLN
ORIGCF806419
Anti-LPIN1 Mouse Monoclonal Antibody [clone: 5H2]
Przeciwciała
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LPIN1
Clonality: Monoclonal
Clone: 5H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR