Drukuj

Anti-ANGPT2 Mouse Monoclonal Antibody [clone: 4A3]

Dostawca: OriGene
ORIGCF806426EA 3840 PLN
ORIGCF806426
Anti-ANGPT2 Mouse Monoclonal Antibody [clone: 4A3]
Przeciwciała
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: ANGPT2
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR