Drukuj

Anti-LPIN1 Mouse Monoclonal Antibody [clone: 5B5]

Dostawca: OriGene
ORIGCF806467EA 3950 PLN
ORIGCF806467
Anti-LPIN1 Mouse Monoclonal Antibody [clone: 5B5]
Przeciwciała
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: LPIN1
Clonality: Monoclonal
Clone: 5B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR