Drukuj

Anti-GABRA5 Mouse Monoclonal Antibody [clone: 1G9]

Dostawca: OriGene
ORIGCF806473EA 3950 PLN
ORIGCF806473
Anti-GABRA5 Mouse Monoclonal Antibody [clone: 1G9]
Przeciwciała
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GABRA5
Clonality: Monoclonal
Clone: 1G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR