Drukuj
Anti-NARS2 Mouse Monoclonal Antibody [clone: 10G5]
Dostawca: OriGene
ORIGCF806498EA 3840 PLN
ORIGCF806498
Anti-NARS2 Mouse Monoclonal Antibody [clone: 10G5]
Przeciwciała
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: NARS2
Clonality: Monoclonal
Clone: 10G5
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human