Drukuj

Anti-NARS2 Mouse Monoclonal Antibody [clone: 10E2]

Dostawca: OriGene
ORIGCF806499EA 3840 PLN
ORIGCF806499
Anti-NARS2 Mouse Monoclonal Antibody [clone: 10E2]
Przeciwciała
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: NARS2
Clonality: Monoclonal
Clone: 10E2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR