Drukuj

Anti-BNIP2 Mouse Monoclonal Antibody [clone: 1C12]

Dostawca: OriGene
ORIGCF806500EA 3840 PLN
ORIGCF806500
Anti-BNIP2 Mouse Monoclonal Antibody [clone: 1C12]
Przeciwciała
Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: BNIP2
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR