Drukuj

Anti-BHLHE41 Mouse Monoclonal Antibody [clone: 8A8]

Dostawca: OriGene
ORIGCF806509EA 3840 PLN
ORIGCF806509
Anti-BHLHE41 Mouse Monoclonal Antibody [clone: 8A8]
Przeciwciała
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150

Type: Primary
Antigen: BHLHE41
Clonality: Monoclonal
Clone: 8A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR