Drukuj

Anti-ATR Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA319394EA 4320 PLN
ORIGTA319394
Anti-ATR Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:15000-1:70000; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: ATR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Fish, Human, Monkey, Mouse, Rat, Xenopus
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR