Drukuj

Anti-CUL2 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA319397EA 4320 PLN
ORIGTA319397
Anti-CUL2 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:2000-1:10000; Western Blot: 1:500-1:1000

Type: Primary
Antigen: CUL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR