Drukuj

Anti-NOTCH2 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA319403EA 4580 PLN
ORIGTA319403
Anti-NOTCH2 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:30000-1:90000; Western Blot: 1:400-1:2000; Immunochemistry: 1:200-1:800

Type: Primary
Antigen: NOTCH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chimpanzee, Dog, Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR