Drukuj

Anti-STAT1 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA319404EA 4320 PLN
ORIGTA319404
Anti-STAT1 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: STAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR