Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-AP2A Goat Polyclonal Antibody

Dostawca: OriGene
Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:2000; Immunoprecipitation: 1:100

Type: Primary
Antigen: AP2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken, Chimpanzee, Dog, Human, Mouse, orangutan, Rat



 

ZAMÓW

Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
100 μg 100 µG ORIGTA319408
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol AP2A
Antigen name adaptor-related protein complex 2, alpha 1 subunit
Reactivity Human, Dog, Chicken, Chimpanzee, Mouse, Orangutan, Rat
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Goat
Western blot Yes
Cross adsorption No
Immunogen This affinity purified antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding aa 3-14 of Human AP2A1 and AP2A2 proteins.
Form Liquid
Storage buffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
Temperatura przechowywania – 20 °C
Shipping temperature –20 °C
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość