Drukuj

Anti-ECT2 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA319414EA 4320 PLN
ORIGTA319414
Anti-ECT2 Rabbit Polyclonal Antibody
Przeciwciała
Recommended Dilutions: ELISA: 1:10000-1:40000; Western Blot: 1:1000-1:4000

Type: Primary
Antigen: ECT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Chimpanzee, Dog, Human, Mouse, Rat, zebraFish
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR