Drukuj

Anti-IL13 Rabbit Polyclonal Antibody

Dostawca: OriGene
ORIGTA328231EA 2830 PLN
ORIGTA328231
Anti-IL13 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: IL13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR