Drukuj

Anti-sII0408 Rabbit Polyclonal Antibody

AGRIAS101594EA 0 PLN
AGRIAS101594
Anti-sII0408 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: SII0408
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Synechocystis sp. pcc 6803
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR