Drukuj
Anti-Hellebrin Rabbit Polyclonal Antibody
AGRIAS101597EA 0 PLN
AGRIAS101597
Anti-Hellebrin Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: Hellebrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: T. aestivum