Drukuj

Anti-Hik29 Rabbit Polyclonal Antibody

AGRIAS101599EA 1520 PLN
AGRIAS101599
Anti-Hik29 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: HIK29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Synechocystis sp. pcc 6803
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR