Drukuj

Anti-AcK Mouse Monoclonal Antibody [clone: 7F8]

Dostawca: Agrisera
AGRIAS10707-25EA 1190 PLN
AGRIAS10707-25
Anti-AcK Mouse Monoclonal Antibody [clone: 7F8]
Przeciwciała
Type: Primary
Antigen: AcK
Clonality: Monoclonal
Clone: 7F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: A. thaliana
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR