Drukuj

Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody [clone: 15A3]

Dostawca: Agrisera
AGRIAS10708-25EA 1190 PLN
AGRIAS10708-25
Anti-8-Hydroxyguanosine Mouse Monoclonal Antibody [clone: 15A3]
Przeciwciała
Type: Primary
Antigen: 8OHDG
Clonality: Monoclonal
Clone: 15A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: A. thaliana
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR