Drukuj

Anti-ICL Chicken Polyclonal Antibody

AGRIAS10713EA 1920 PLN
AGRIAS10713
Anti-ICL Chicken Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: ICL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Streptomyces sp.
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR