Drukuj

Anti-Ramy 3D Rabbit Polyclonal Antibody

Dostawca: Agrisera
AGRIAS10716EA 2250 PLN
AGRIAS10716
Anti-Ramy 3D Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: RAMY 3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Oryza sativa
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR