Drukuj

Anti-PHOT1 Rabbit Polyclonal Antibody

Dostawca: Agrisera
AGRIAS10720EA 1600 PLN
AGRIAS10720
Anti-PHOT1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: PHOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR