Anti-PEPC Rabbit Polyclonal Antibody
ROCK200-4163-0100
- Antibody type:Primary
- Antigen symbol:PEPC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Maize
- Western blot:Yes
- Cross adsorption:No
- Gene ID:542372
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Anti-Phospho Enol Pyruvate Carboxylase Antibody was produced by repeated immunizations with maize leaves Phospho-enol-pyruvate Carboxylase.
- Purification:purified by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer
- Size:100 µg
- Pk:100 µG
Anti-Phospho Enol Pyruvate Carboxylase antibody is suitable for western blotting, IHC and for ELISA. Researchers should determine optimal titers for applications that are not stated below.
Recommended Dilutions: ELISA: 1:10000 - 1:40000; Western Blot: 1:1000 - 1:4000; Immunochemistry: User Optimized
Type: Primary
Antigen: PEPC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Maize Leaves, Maize