Anti-LOX Rabbit Polyclonal Antibody
ROCK100-401-H91
- Antibody type:Primary
- Antigen name:lysyl oxidase
- Antigen symbol:LOX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:internal
- Cross adsorption:No
- Gene ID:4015
- Storage buffer:0,05% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Lysyl Oxidase Antibody was produced from whole rabbit serum prepared by repeated immunizations with the human protein Lysyl Oxidase at an internal amino acid sequence.
- Purification:Anti-Lysyl Oxidase Antibody is crude rabbit sera. A BLAST analysis was used to suggest cross-reactivity with Anti-Lysyl Oxidase from human, bovine, chimpanzee, mouse and rat 100% homology with the immunizing sequence. Predicted reactivity with chicken, dog, pig, and xenopus. Cross-reactivity with Anti-Lysyl Oxidase from other sources has not been determined.
- Size:100 µl
- Pk:100 µl
Anti-Lysyl Oxidase antibody is suitbale for use in IF/ICC, IHC (frozen), IHC (p), IP and WB. Expect a band approximately 31kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Western Blot: 1:250-1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: LOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse