Peha-Soft® Nitrile White Powderfree Nitrile Gloves
Supplier: HARTMANN PAUL
Certificates
About this item
These latex-free disposable examination and protective gloves are made of soft nitrile rubber.
- With chlorinated inner surface for easy donning
- Texturing of the finger area ensures optimum grip
- Highly elastic and especially tear-resistant
- Excellent fit and high tactile sensitivity
- Can be worn on either hand
Ideal for use in all areas where staff are at particular risk of contamination; especially suitable for persons prone to latex allergies or who experience hypersensitivity reactions to glove powder. Also applicable in laboratories e.g. for contact with chemicals and disinfectants. May be used in direct contact with food.
Certifications: EN 374, EN 455, Cat. I+II. Food contact approved.
Specifications
- Catalog No:
- HART9422082
- HART9422092
- HART9422063
- HART9422181
- HART9422072
- HART9422161
- HART9422171
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- EN 455, EN 374
- Food contact approved:
- +
- +
- +
- +
- +
- +
- +
Specifications
Frequently Bought Together