CF™543 antibodies are affinity-purified antibodies labeled with orange fluorescent CF™543 (Ex/Em 541/560nm), a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™543 is an orange fluorescent dye with an excitation spectrum optimally matching the 543 laser line. Antibody conjugates of the dye are much brighter than those of Alexa Fluor® 546.
CF™ dyes are covered by pending U.S. and international patents.
Informacje do zamówienia: Antibodies are supplied at 2mg/mL in PBS pH 7.4 containing 50% glycerol, 2mg/mL BSA (IgG- and protease-free) and 0.05% sodium azide. Highly cross-adsorbed antibodies are adsorbed against whole serum protein and have minimal cross-reactivity for multiple labeling experiments as indicated. Bv: bovine; Ch: Chicken; Gt: goat; GP: Guinea pig; Hs: horse; Hu: human; Ms: mouse; Rb: rabbit; Sh: sheep; SHm: Syrian hamster; Sw: swine; Rt: rat.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF543
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Goat