Drukuj

Anti-HIF1A Rabbit Polyclonal Antibody

ABNOPAB7981EA 1820 PLN
ABNOPAB7981
Anti-HIF1A Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against synthetic peptide of HIF1A.

Recommended Dilutions: Western Blot: 2 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1-2 ?g/ml

Type: Primary
Antigen: HIF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rabbit, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR