Drukuj

Anti-CHRNA4 Goat Polyclonal Antibody

Dostawca: Abnova
ABNOPAB7534EA 2040 PLN
ABNOPAB7534
Anti-CHRNA4 Goat Polyclonal Antibody
Przeciwciała
Goat polyclonal antibody raised against synthetic peptide of CHRNA4.

Recommended Dilutions: ELISA: 1:4000

Type: Primary
Antigen: CHRNA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR