Drukuj

Anti-EFR3A Rabbit Polyclonal Antibody

Dostawca: Abnova
ABNOPAB21532EA 2650 PLN
ABNOPAB21532
Anti-EFR3A Rabbit Polyclonal Antibody
Przeciwciała
Rabbit polyclonal antibody raised against recombinant EFR3A.

Recommended Dilutions: Immunohistochemistry: 1:50-1:200; Western Blot: 1:250-1:500

Type: Primary
Antigen: EFR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR