Drukuj
Zestaw testowy MASTASTAPH
710-0213EA 946 PLN
MAST181011
Zestaw testowy MASTASTAPH
For the identification of Staphylococci, which synthezise the Clumping factor and/or Protein A.
  • For 120 tests