Drukuj
Drabinka DNA, 100 bp, DNAmark™
Dostawca: G-Biosciences
GENO786-855EA 648 PLN
GENO786-855
Drabinka DNA, 100 bp, DNAmark™
Elektroforezy, Odczynniki Markery masy cząsteczkowej
DNAmark™ 100 bp ladder consists of 11 fragments ranging from 100 to 1500 base pairs (bp). The 500 bp fragment is present at increased intensity to allow easy identification. All fragments are precisely quantified and mixed during the production. For 5 µl loading, all fragments except 500 bp are 40 ng. The 500 bp fragment is 100 ng.
  • Pre-mixed with loading dye and ready to use