Drukuj

Storage boxes

Dostawca: Multiroir
MULC41743EA 5140 PLN
MULC41743
Storage boxes
PS, crystal.

  • Hook closure to secure contents
  • Removable dimpled white grids
  • Transparent hinged lid

These boxes with transparent cover help in high visibility of it's contents.

Colour: Blue
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR