The ImmunoWash™ 1575 microplate washer is essential for performing accurate and reproducible immunoassays in a microplate format, such as ELISA. It is suitable for autoclave sterilisation and compatible with 96-well microplates that have flat-, U-, or V-bottom wells and strips.
- Choose from a variety of preprogrammed wash sequences
- Design, store, and reproducibly perform special wash routines
- Maximise wash efficiency by adapting to various liquids and buffers, assay components and conditions
- Exercise complete control of needle position (horizontal and vertical) and aspiration/dispensing rates
Dimensions: 345×423×200 mm
Weight: 10,3 kg