Insoles, uvex 1 climate comfort
Supplier: UVEX
Certificates
About this item
These comfortable insoles are made of highly breathable and moisture absorbent material.
- Perforation for improved breathability
- Exceptional, full-surface shock absorption ensures comfort in high-strain zone
- Non irritating, comfortable surface structure and anti-static properties
Exceptional, full-surface shock absorption ensures comfort in high-strain zones.
Colour: Black/red
Width: 10
Width: 10
Product Information
- Catalog #Supplier #Shoe sizeColourPkAvailabilityPrice
- 114-61199534.7/4444Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61209534.7/4545Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61219534.7/4646Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61319534.7/51-5233Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61309534.7/49-5049/50Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61139534.7/3535Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61279534.7/3636Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61289534.7/3737Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61149534.7/3838Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61159534.7/3939Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61169534.7/4040Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61299534.7/4141Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61179534.7/4242Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61189534.7/4343Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61229534.7/4747Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-61239534.7/4848Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable
Width: 11
Product Information
- Catalog #Supplier #Shoe sizeColourPkAvailabilityPrice
- 114-74699534.8/3535Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74809534.8/4646Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74839534.8/49-5049/50Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74849534.8/51-5251/52Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74759534.8/4141Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74789534.8/4444Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74819534.8/4747Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74769534.8/4242Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74779534.8/4343Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74799534.8/4545Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74829534.8/4848Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74719534.8/3737Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74709534.8/3636Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74729534.8/3838Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74739534.8/3939Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74749534.8/4040Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable
Width: 12
Product Information
- Catalog #Supplier #Shoe sizeColourPkAvailabilityPrice
- 114-74989534.9/4848Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74859534.9/3535Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74869534.9/3636Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74969534.9/4646Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74979534.9/4747Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74999534.9/49-5049/50Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75009534.9/51-5251/52Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74879534.9/3737Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74889534.9/3838Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74899534.9/3939Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74909534.9/4040Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74919534.9/4141Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74929534.9/4242Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74939534.9/4343Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74949534.9/4444Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-74959534.9/4545Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable
Width: 14
Product Information
- Catalog #Supplier #Shoe sizeColourPkAvailabilityPrice
- 114-75039534.0/3737Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75049534.0/3838Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75099534.0/4343Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75019534.0/3535Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75139534.0/4747Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75149534.0/4848Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75029534.0/3636Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75089534.0/4242Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75169534.0/51-5251/52Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75159534.0/49-5049/50Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75109534.0/4444Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75119534.0/4545Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75129534.0/4646Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75079534.0/4141Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75059534.0/3939Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable - 114-75069534.0/4040Black/red1 PAIR1 * 1 PARA97,50 złSpecifications:
Specifications
Environmentally Preferable