Drukuj
safesolv solvent substitute, Q Path® do histologii, for pathology
Dostawca: VWR Q-Path Chemicals
Niebezpieczeństwo
00699464.EA 357 PLN
00699464.
safesolv solvent substitute, Q Path® do histologii, for pathology
safesolv
Q Path® Safesolv is a non hazardous replacement for xylene or toluene. It is compatible with the Q Path® Safemount mounting medium reference 00647520. It is used in place of xylene, toluene, methylecyclohexane or other solvents in the course of the dehydration stages of histological specimens and during the stages of staining and lamella mounting.
Jednostka: 3295
ADR: 3,III
IVDLogo
Zobacz ceny
Wyniki testów specyfikacyjnych
Appearance Clear colourless liquid
Aromatics (UV) (max. 100 ppm) Passes test
Boiling point (begin) (min.173°C) Passes test
Bromine (per 100 g) (max. 50 ppm) Passes test
Colouration (Saybolt) (D6045) (min. 30) Passes test
Density (15°C) (0.752-0.772 kg/l) Passes test
Distillation range (50 %) (177-187°C) Passes test
Dry point (boiling range) (max.193°C) Passes test
Flash point (56-63°C) Passes test
S (Sulphur) (max. 1 ppm) Passes test