Tissue DNA Mini kits for the quick and reliable isolation of high quality DNA from a wide range of animal and human cells and tissue. An optimised MicroSpin option is available for very small sample amounts.
- No need for organic extractions
- No need for time-consuming alcohol precipitations
- Total enzymatic degradation of cellular RNA
- Efficient separation of enzyme inhibitors and contaminants
- Final elution with low salt buffer or water
*Safety Line columns with snap-on lids, Classic Line columns without lids
Dodatkowe informacje: Kits contain either PerfectBind DNA columns (Safety or Classic Line*) or PerfectBind MicroSpin DNA columns. All kits contain collection tubes, DNA Lysis Buffer T, DNA Binding Buffer, DNA Wash Buffer, TE Buffer, Elution Buffer and Proteinase K.