Drukuj
Qualitative filter papers, technical, smooth, grade 10
Dostawca: Sartorius
SARTFT-3-352-110EA 29.9 PLN
SARTFT-3-352-110 SARTFT-3-352-055 SARTFT-3-352-240 SARTFT-3-352-185 SARTFT-3-352-125 SARTFT-4-352-270 SARTFT-3-352-070 SARTFT-3-352-090 SARTFT2-352-580580 SARTFT-3-352-150
Qualitative filter papers, technical, smooth, grade 10
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%.
They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Fast rate of filtration, wide pore.