Anti-GABRB3 Antibody
BSBTPB9595
- Antibody type:Primary
- Antigen name:gamma-aminobutyric acid (GABA) A receptor, beta 3
- Antigen symbol:GABRB3
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2562
- Antigen synonyms:ECA5
- Size:100 µg
- Pk:100 µG
Rabbit IgG polyclonal antibody for Gamma-aminobutyric acid receptor subunit beta-3(GABRB3) detection. Tested with WB in Human;Mouse; Rat.
Type: Primary
Antigen: GABRB3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human