Drukuj
Drukarki etykiet, BBP™35 i BBP™37
Dostawca: Brady
BRDY198560EA 20830 PLN
BRDY198560 BRDY198562 116-0596 116-0595 116-0598 116-0597 116-0600 116-0599 116-0602 116-0601
Drukarki etykiet, BBP™35 i BBP™37
Drukarki specjalistyczne Label Printers
The BBP™35 and BBP™37 sign and label printers with 300 dpi print resolution can produce signs and labels in the colours and shapes that can make a strong visual impact in the workplace.
  • Multiple print colours
  • Automatic label set-up
  • 20 second material change overs, simply 'drop in' the ribbon and labels with no complicated threading or assembly
  • 12,7 cm/second print speed
  • Intuitive built-in menu features for easy operation
  • Maximum label width: 101 mm
BBP™37 additional features.
Shape cutting with built-in XY plotter-cutter system
Cut out shapes and text or create message labels to fit the user requirements
Dodatkowe informacje: Printer is supplied with power cord, USB cable, cleaning kit, cutter cleaning tool, and quick start guide.