Poland
Order Entry
Anti-HBcAg Rabbit Polyclonal Antibody
Anti-HBcAg Rabbit Polyclonal Antibody
  BOSSBS-15455R
 :  Bioss
Anti-HBcAg Rabbit Polyclonal Antibody
  BOSSBS-15455R
 :  Bioss
 
5 470 zł
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen symbol:
    HBcAg
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Virus
  • Western blot:
    Yes
  • Form:
    liquid
  • Antigen synonyms:
    C|HBc|Core and e antigen|Hepatitis B Virus core antigen|Core protein|Capsid protein|CAPSD_HBVA3.|Core antigen|precore/core protein|HBVgp4|p21.5|HBcAg
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Hepatitis B Virus Core Antigen (HBcAg) is part of the infectious virion containing an inner "core particle" enclosing the viral genome. The icosahedral core particle contains 180 or 240 copies of the core protein. HBcAg is one of the three major clinical antigens of hepatitis B virus but disappears early in the course of infection.The hepatitis B virus core antigen (HBcAg) is a highly immunogenic subviral particle and functions as both a T-cell-dependent and a T-cell-independent antigen. Therefore, HBcAg may be a promising candidate target for therapeutic vaccine control of chronic HBV infection.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: HBcAg
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus