Drukuj
Źródła zimnego światła LED, duża moc, VisiLight® LED7
631-1105EA 6140 PLN
VWRI631-1105 VWRI631-1102
Źródła zimnego światła LED, duża moc, VisiLight® LED7
These high power VisiLight® LED cold light sources are characterised by superior finishing quality. Due to the environmentally friendly LED technology they are durable and perform efficiently. The LED used in these cold light sources are particularly powerful, and are the perfect choice for lighting applications where high or extreme brightness is required. Expected LED lifespan of 50000 hours.
  • Modern design with rugged anodised aluminium housing
  • Vertical and horizontal positioning, stackable
  • Environmentally friendly LED technology with flicker-free illumination
  • LED7 provides very high brightness (>1300 lm)
  • Glare protection by light guide detection
W×D×H: 174×259×137 mm
Weight: 2,7 kg
* Light guide of active Ø = 9 mm and L = 1 m