About this item
These compact bench top tube furnaces with integrated controllers can be widely used for a variety of processes. Equipped with a standard C 530 ceramic working tube and two fibre plugs they have an unbeatable price/performance ratio. Units feature rust-free, textured stainless steel exteriors. R models feature Tmax 1200 or 1300 °C, RD models have Tmax 1100 or 1300 °C.
- Tube diameter: RD models 30 mm, R models 50 to 170 mm
- Heated length: RD models 200 mm, R models 250 to 1000 mm
- C 530 ceramic working tube and two fibre plugs supplied as standard
- R and RD models feature S type thermocouple, except RD model with Tmax 1100 which has a K type thermocouple
- Silent solid state power control relay
- Non-classified insulation avoids the potential risk of cancer
- Controller B 510 and C 550 can be read out via an integrated USB-process documentation
* ΔT 10 °C, uniform length measured with end plugs fitted.
** 380 V three phase power supply.
*** ΔT 5 °C, uniform length measured with end plugs fitted.
RD series, Tmax 1100 °C model with R 7 controller with one storable holding temperature
Product Information
- Catalog #Supplier #TypeMax. temp.PowerØ int. tubeTube lengthHeated lengthAvailabilityPrice
- 455-0487RD30A171ARD 30/200/111100 °C1500 W30 mm~340 mm200 mmLog in for availability1 * 1 SZT18 890 złSpecifications:
Specifications
Environmentally Preferable
R series, Tmax 1200 °C models with Standard B 510 controller with 5 programmes and 4 segments
Product Information
- Catalog #Supplier #TypeMax. temp.PowerØ int. tubeTube lengthHeated lengthAvailabilityPrice
- 455-2064R-152L2ANR 50/250/121200 °C1600 W40 mm450 mm250 mmLog in for availability1 * 1 SZT22 840 złSpecifications:
Specifications
Environmentally Preferable - 455-2074R-322L2ANR 120/500/12**1200 °C6500 W100 mm850 mm500 mmLog in for availability1 * 1 SZT32 370 złSpecifications:
Specifications
Environmentally Preferable - 455-2080R-472L2ANR 170/750/12**1200 °C10000 W150 mm1100 mm750 mmLog in for availability1 * 1 SZT49 130 złSpecifications:
Specifications
Environmentally Preferable - 455-2086R-572L2ANR 170/1000/12**1200 °C11500 W150 mm1350 mm1000 mmLog in for availability1 * 1 SZT57 990 złSpecifications:
Specifications
Environmentally Preferable - 455-2068R-252L2ANR 50/500/121200 °C2300 W40 mm700 mm500 mmLog in for availability1 * 1 SZT25 960 złSpecifications:
Specifications
Environmentally Preferable
R series, Tmax 1300 °C models with Standard B 510 controller with 5 programmes and 4 segments
Product Information
- Catalog #Supplier #TypeMax. temp.PowerØ int. tubeTube lengthHeated lengthAvailabilityPrice
- 455-2065R-152L3ANR 50/250/131300 °C1600 W40 mm450 mm250 mmLog in for availability1 * 1 SZT24 460 złSpecifications:
Specifications
Environmentally Preferable - 455-2087R-572L3ANR 170/1000/13**1300 °C11500 W150 mm1350 mm1000 mmLog in for availability1 * 1 SZT66 120 złSpecifications:
Specifications
Environmentally Preferable - 455-2081R-472L3ANR 170/750/13**1300 °C10000 W150 mm1100 mm750 mmLog in for availability1 * 1 SZT55 590 złSpecifications:
Specifications
Environmentally Preferable - 455-2075R-322L3ANR 120/500/13**1300 °C6500 W100 mm850 mm500 mmLog in for availability1 * 1 SZT36 770 złSpecifications:
Specifications
Environmentally Preferable - 455-2069R-252L3ANR 50/500/131300 °C2300 W40 mm700 mm500 mmLog in for availability1 * 1 SZT28 750 złSpecifications:
Specifications
Environmentally Preferable
R series, Tmax 1200 °C models with Professional C 550 controller with 10 programmes and 20 segments
Product Information
- Catalog #Supplier #TypeMax. temp.PowerØ int. tubeTube lengthHeated lengthAvailabilityPrice
- 455-2066R-152O2ANR 50/250/121200 °C1600 W40 mm450 mm250 mmLog in for availability1 * 1 SZT27 130 złSpecifications:
Specifications
Environmentally Preferable - 455-2088R-572O2ANR 170/1000/12**1200 °C11500 W150 mm1350 mm1000 mmLog in for availability1 * 1 SZT62 280 złSpecifications:
Specifications
Environmentally Preferable - 455-2082R-472O2ANR 170/750/12**1200 °C10000 W150 mm1100 mm750 mmLog in for availability1 * 1 SZT53 420 złSpecifications:
Specifications
Environmentally Preferable - 455-2076R-322O2ANR 120/500/12**1200 °C6500 W100 mm850 mm500 mmLog in for availability1 * 1 SZT36 660 złSpecifications:
Specifications
Environmentally Preferable - 455-2070R-252O2ANR 50/500/121200 °C2300 W40 mm700 mm500 mmLog in for availability1 * 1 SZT30 250 złSpecifications:
Specifications
Environmentally Preferable
R series, Tmax 1300 °C models with Professional C 550 controller with 10 programmes and 20 segments
Product Information
- Catalog #Supplier #TypeMax. temp.PowerØ int. tubeTube lengthHeated lengthAvailabilityPrice
- 455-2083R-472O3ANR 170/750/13**1300 °C10000 W150 mm1100 mm750 mmLog in for availability1 * 1 SZT59 880 złSpecifications:
Specifications
Environmentally Preferable - 455-2077R-322O3ANR 120/500/13**1300 °C6500 W100 mm850 mm500 mmLog in for availability1 * 1 SZT41 060 złSpecifications:
Specifications
Environmentally Preferable - 455-2067R-152O3ANR 50/250/131300 °C1600 W40 mm450 mm250 mmLog in for availability1 * 1 SZT28 750 złSpecifications:
Specifications
Environmentally Preferable - 455-2089R-572O3ANR 170/1000/13**1300 °C11500 W150 mm1350 mm1000 mmLog in for availability1 * 1 SZT70 410 złSpecifications:
Specifications
Environmentally Preferable - 455-2071R-252O3ANR 50/500/131300 °C2300 W40 mm700 mm500 mmLog in for availability1 * 1 SZT33 040 złSpecifications:
Specifications
Environmentally Preferable
R series, Tmax 1200 °C models with Professional P 580 controller with 50 programmes and 40 segments
Product Information
- Catalog #Supplier #TypeMax. temp.PowerØ int. tubeTube lengthHeated lengthAvailabilityPrice
- 455-2084R-472W2AN3R 170/750/12**1200 °C10000 W150 mm1100 mm750 mmLog in for availability1 * 1 SZT56 820 złSpecifications:
Specifications
Environmentally Preferable - 455-2078R-322W2AN3R 120/500/12**1200 °C6500 W100 mm850 mm500 mmLog in for availability1 * 1 SZT40 050 złSpecifications:
Specifications
Environmentally Preferable - 455-2090R-572W2AN3R 170/1000/12**1200 °C11500 W150 mm1350 mm1000 mmLog in for availability1 * 1 SZT65 680 złSpecifications:
Specifications
Environmentally Preferable - 455-2072R-252W2AN3R 50/500/121200 °C2300 W40 mm700 mm500 mmLog in for availability1 * 1 SZT33 650 złSpecifications:
Specifications
Environmentally Preferable
R series, Tmax 1300 °C models with Professional P 580 controller with 50 programmes and 40 segments
Product Information
- Catalog #Supplier #TypeMax. temp.PowerØ int. tubeTube lengthHeated lengthAvailabilityPrice
- 455-2085R-472W3AN3R 170/750/13**1300 °C10000 W150 mm1100 mm750 mmLog in for availability1 * 1 SZT63 280 złSpecifications:
Specifications
Environmentally Preferable - 455-2079R-322W3AN3R 120/500/13**1300 °C6500 W100 mm850 mm500 mmLog in for availability1 * 1 SZT44 450 złSpecifications:
Specifications
Environmentally Preferable - 455-2091R-572W3AN3R 170/1000/13**1300 °C11500 W150 mm1350 mm1000 mmLog in for availability1 * 1 SZT73 810 złSpecifications:
Specifications
Environmentally Preferable - 455-2073R-252W3AN3R 50/500/131300 °C2300 W40 mm700 mm500 mmLog in for availability1 * 1 SZT36 430 złSpecifications:
Specifications
Environmentally Preferable