Columns, HiTrap™ Chelating Sepharose FF
Supplier: Cytiva
Certificates
Specifications
- Catalog No:
- GEHE90-1004-80
- 90-1003-47
- Storage:
- 4 to 30 °C, 20% Ethanol
- 4 to 30 °C, 20% Ethanol
Specifications
- Column hardwareParticle SizeBed dimensionsCapacityPk
- HiTrap™ Chelating Sepharose FFHiTrap™ Chelating Sepharose FF90-1003-4790-1003-47PP90 µm7×25 mm1 ml51 * 5 SZT3 730 zł: 90-1003-47: GEHE90-1003-47
Specifications
Environmentally PreferableStorage4 to 30 °C, 20% Ethanol - HiTrap™ Chelating Sepharose FFHiTrap™ Chelating Sepharose FFGEHE90-1004-8090-1004-80PP90 µm16×25 mm5 ml51 * 5 SZT7 260 zł: 90-1004-80: GEHE90-1004-80
Specifications
Environmentally PreferableStorage4 to 30 °C, 20% Ethanol