Keep my session open?
Ending In 
The session is expired
Twoja sesja wygasła. Zostałeś wylogowany dla własnego bezpieczeństwa.
Chcesz zalogować się jeszcze raz?

Anti-SUV39H2 Rabbit Polyclonal Antibody

Dostawca: Abgent
Type: Primary
Antigen: SUV39H2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human




 

ZAMÓW

Rozwiń / Zwiń
Ilość Numer katalogowy VWR Ilość
400 µl ABGEAP19115B
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF

SPECYFIKACJE

Antigen symbol SUV39H2
Antigen name suppressor of variegation 3-9 homolog 2 (Drosophila)
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
Antigen synonyms KMT1B
Amino acid number 367 to 395
Gene ID 79723
Epitope C-terminal
Stężenie 0.65
Immunogen This SUV39H2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 367-395 amino acids from the C-terminal region of human SUV39H2.
Molecular weight 46.682 kDa
Purification Protein A & Peptide affinity purification
Sequence SDSIDHSPAKKRVRC
Storage buffer pH 7,4
Temperatura przechowywania Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Wprowadź szczegóły

Select the article for which you are looking for Certificate

Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość