Chcesz zalogować się jeszcze raz?
Anti-ASB7 Rabbit Polyclonal Antibody
Dostawca: Abgent
Type: Primary
Antigen: ASB7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
ZAMÓW
Rozwiń / Zwiń
Ilość Numer katalogowy VWR Ilość
400 µl ABGEAP18060C
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
SPECYFIKACJE
Antigen symbol ASB7
Antigen name ankyrin repeat and SOCS box containing 7
Reactivity Human
Conjugation Unconjugated
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Flow cytometry Yes
Amino acid number 82 to 108
Gene ID 140460
Epitope internal
Stężenie 0.34
Immunogen This ASB7 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 82-108 amino acids from the Central region of human ASB7.
Molecular weight 36.011 kDa
Purification Protein A & Peptide affinity purification
Sequence ARLMLESEYRSDC
Storage buffer pH 7,4
Temperatura przechowywania Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at – 20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość