Drukuj

Anti-BICC1 Rabbit Polyclonal Antibody

Dostawca: Abgent
ABGEAP20561AEA 1750 PLN
ABGEAP20561A
Anti-BICC1 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: BICC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR