Drukuj
Anti-CCDC134 Rabbit Polyclonal Antibody
Dostawca: Abgent
ABGEAP13242BEA 1750 PLN
ABGEAP13242B
Anti-CCDC134 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: CCDC134
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human