Drukuj

Anti-RPAP3 Rabbit Polyclonal Antibody

Dostawca: Abgent
ABGEAP18284CEA 1750 PLN
ABGEAP18284C
Anti-RPAP3 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: RPAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR