Drukuj
Anti-RANBP3 Rabbit Polyclonal Antibody
Dostawca: Abgent
ABGEAP16369AEA 1750 PLN
ABGEAP16369A
Anti-RANBP3 Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: RANBP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human